Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280707.1 | NP_001267636.1 | 4 - 5 | 558 | 101 | |
XM_015102519.2 | XP_014958005.2 | 5 - 6 | 604 | 101 | ||
GenBank mRNA | GU585526.1 | - | 4 - 5 | 558 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |