Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097578.2 | XP_014953064.2 | 1 - 2 | 170 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |