Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139450.1 | NP_001132922.1 | 1 - 2 | 115 | 82 | |
GenBank mRNA | EU293836.1 | - | 1 - 2 | 115 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |