Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097262.2 | XP_014952748.2 | 2 - 3 | 271 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |