Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958652.1 | XP_027814453.1 | 10 - 11 | 1206 | 60 | |
XM_027958653.1 | XP_027814454.1 | 10 - 11 | 978 | 60 | ||
XM_027958654.1 | XP_027814455.1 | 7 - 8 | 773 | 60 | ||
XM_027958655.1 | XP_027814456.1 | 10 - 11 | 1212 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |