Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958652.1 | XP_027814453.1 | 11 - 12 | 1342 | 69 | |
XM_027958653.1 | XP_027814454.1 | 11 - 12 | 1114 | 69 | ||
XM_027958654.1 | XP_027814455.1 | 8 - 9 | 909 | 69 | ||
XM_027958655.1 | XP_027814456.1 | 11 - 12 | 1348 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |