Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968694.1 | XP_027824495.1 | 3 - 4 | 458 | 73 | |
XM_027968695.1 | XP_027824496.1 | 5 - 6 | 594 | 73 | ||
XM_027968696.1 | XP_027824497.1 | 6 - 7 | 651 | 73 | ||
XM_027968697.1 | XP_027824498.1 | 5 - 6 | 589 | 73 | ||
XM_027968698.1 | XP_027824499.1 | 4 - 5 | 501 | 73 | ||
XM_027968699.1 | XP_027824500.1 | 2 - 3 | 380 | 73 | ||
XM_027968701.1 | XP_027824502.1 | 3 - 4 | 445 | 73 |