Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114667.1 | NP_001108139.1 | 2 - 3 | 241 | 80 | |
GenBank mRNA | EU301804.1 | - | 2 - 3 | 241 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |