Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161862.1 | NP_001155334.1 | 3 - 4 | 194 | 120 | |
GenBank mRNA | FJ943970.1 | - | 3 - 4 | 194 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |