Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126368.1 | NP_001119840.1 | 4 - 5 | 466 | 95 | |
XM_012182368.3 | XP_012037758.1 | 5 - 6 | 713 | 95 | ||
GenBank mRNA | EU016225.1 | - | 4 - 5 | 466 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |