Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009201.1 | NP_001009201.1 | 4 - 5 | 679 | 88 | |
XM_015104664.2 | XP_014960150.2 | 5 - 6 | 845 | 88 | ||
GenBank mRNA | AF034842.1 | - | 4 - 5 | 679 | 88 |