Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134306.1 | NP_001127778.1 | 2 - 3 | 485 | 111 | |
XM_027965721.1 | XP_027821522.1 | 4 - 5 | 889 | 111 | ||
XM_027965722.1 | XP_027821523.1 | 4 - 5 | 667 | 111 | ||
XM_027965723.1 | XP_027821524.1 | 4 - 5 | 733 | 111 | ||
GenBank mRNA | EU797605.1 | - | 2 - 3 | 485 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |