Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306115.1 | NP_001293044.1 | 3 - 4 | 505 | 65 | |
XM_004018278.3 | XP_004018327.1 | 3 - 4 | 1402 | 65 | ||
XM_012099534.3 | XP_011954924.1 | 3 - 4 | 662 | 65 | ||
XM_012099535.3 | XP_011954925.1 | 3 - 4 | 255 | 65 | ||
GenBank mRNA | KM199684.1 | - | 3 - 4 | 505 | 65 |