Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077209.2 | NP_001070677.2 | 2 | 1066 | 73 | |
GenBank mRNA | AM117197.1 | - | 1 | 569 | 73 | |
DQ917485.1 | - | 1 | 920 | 73 | ||
KY110723.1 | - | 1 | 920 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |