Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139449.1 | NP_001132921.1 | 1 - 2 | 144 | 61 | |
GenBank mRNA | EU293835.1 | - | 1 - 2 | 144 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |