Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290006.2 | NP_001276935.1 | 3 - 4 | 527 | 125 | |
GenBank mRNA | GT644620.1 | - | 3 - 4 | 551 | 125 | |
GT873426.1 | - | 3 - 4 | 446 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |