Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308575.1 | NP_001295504.1 | 4 - 5 | 454 | 107 | |
XM_027975563.1 | XP_027831364.1 | 5 - 6 | 759 | 107 | ||
XM_027975564.1 | XP_027831365.1 | 4 - 5 | 674 | 107 | ||
GenBank mRNA | HF674450.1 | - | 4 - 5 | 454 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |