Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166201.1 | NP_001159673.1 | 3 - 4 | 1247 | 90 | |
GenBank mRNA | GQ221057.1 | - | 3 - 4 | 1247 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |