Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009415.1 | NP_001009415.1 | 7 - 8 | 1051 | 89 | |
GenBank mRNA | X60149.1 | - | 912 | 89 | ||
Z11560.1 | - | 1051 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |