Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163605.1 | NP_001157077.1 | 4 - 5 | 489 | 92 | |
XM_015092739.2 | XP_014948225.1 | 5 - 6 | 1025 | 92 | ||
XM_027963869.1 | XP_027819670.1 | 5 - 6 | 1109 | 92 | ||
GenBank mRNA | FJ937959.1 | - | 4 - 5 | 489 | 92 |