Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282137.1 | NP_001269066.1 | 3 - 4 | 452 | 59 | |
XM_012095254.3 | XP_011950644.2 | 5 - 6 | 676 | 59 | ||
XM_012095257.3 | XP_011950647.2 | 4 - 5 | 609 | 59 | ||
XM_012095260.3 | XP_011950650.2 | 6 - 7 | 647 | 59 | ||
XM_012095261.3 | XP_011950651.2 | 5 - 6 | 561 | 59 | ||
GenBank mRNA | EE776125.1 | - | 3 - 4 | 452 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |