Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306103.1 | NP_001293032.1 | 1 | 192 | 104 | |
GenBank mRNA | KJ000742.1 | - | 1 | 192 | 104 |
Gene Symbol: LOC114115289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970878.1 | XP_027826679.1 | 1 | 188 | 104 | |
XM_027970879.1 | XP_027826680.1 | 1 | 188 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |