Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306110.1 | NP_001293039.1 | 4 - 5 | 632 | 63 | |
XM_027975558.1 | XP_027831359.1 | 3 - 4 | 3168 | 63 | ||
XM_027975559.1 | XP_027831360.1 | 4 - 5 | 2549 | 63 | ||
GenBank mRNA | KJ159212.1 | - | 4 - 5 | 632 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |