Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009441.1 | NP_001009441.1 | 2 - 3 | 403 | 87 | |
GenBank mRNA | AF092739.1 | - | 2 - 3 | 403 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |