Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127276.1 | NP_001120748.1 | 3 - 4 | 449 | 96 | |
XM_015097954.2 | XP_014953440.1 | 4 - 5 | 630 | 96 | ||
GenBank mRNA | EU626212.1 | - | 3 - 4 | 449 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |