Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127276.1 | NP_001120748.1 | 4 - 5 | 623 | 129 | |
XM_015097954.2 | XP_014953440.1 | 5 - 6 | 804 | 129 | ||
GenBank mRNA | EU626212.1 | - | 4 - 5 | 623 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |