Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127277.1 | NP_001120749.1 | 5 - 6 | 534 | 104 | |
GenBank mRNA | EU626214.1 | - | 5 - 6 | 534 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |