Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126352.1 | NP_001119824.1 | 1 | 115 | 92 | |
XM_015095469.2 | XP_014950955.1 | 3 - 4 | 228 | 92 | ||
XM_015095470.2 | XP_014950956.1 | 3 - 4 | 209 | 92 | ||
XM_027969158.1 | XP_027824959.1 | 3 - 4 | 201 | 92 | ||
GenBank mRNA | EU596596.1 | - | 1 | 115 | 92 |
Gene Symbol: LOC114116592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973508.1 | XP_027829309.1 | 1 | 140 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |