Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009721.3 | NP_001009721.1 | 2 - 3 | 155 | 68 | |
GenBank mRNA | AB060699.1 | - | 1 - 2 | 147 | 68 | |
AY228762.1 | - | 1 - 2 | 68 | 68 | ||
DQ152959.1 | - | 1 - 2 | 93 | 68 | ||
DQ294307.1 | - | 2 - 3 | 159 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |