Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160027.1 | NP_001153499.1 | 1 - 2 | 122 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |