Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009805.1 | NP_001009805.1 | 2 - 3 | 209 | 80 | |
XM_012177585.3 | XP_012032975.1 | 2 - 3 | 370 | 80 | ||
GenBank mRNA | X53561.1 | - | 209 | 80 | ||
X55991.1 | - | 196 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |