Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009805.1 | NP_001009805.1 | 3 - 4 | 341 | 91 | |
GenBank mRNA | X53561.1 | - | 341 | 91 | ||
X55991.1 | - | 328 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |