Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161867.1 | NP_001155339.1 | 2 - 3 | 441 | 119 | |
GenBank mRNA | FJ943975.1 | - | 2 - 3 | 441 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |