Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEGAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009766.1 | NP_001009766.1 | 5 - 6 | 398 | 107 | |
GenBank mRNA | AY509922.1 | - | 4 - 5 | 283 | 107 | |
AY509923.1 | - | 5 - 6 | 373 | 107 | ||
AY509924.1 | - | 4 - 5 | 308 | 107 | ||
AY509925.1 | - | 5 - 6 | 398 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |