Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164713.1 | NP_001158185.1 | 5 - 6 | 437 | 93 | |
GenBank mRNA | FJ039532.1 | - | 5 - 6 | 437 | 93 | |
KJ425421.1 | - | 5 - 6 | 436 | 93 |