Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280694.1 | NP_001267623.1 | 3 - 4 | 745 | 90 | |
GenBank mRNA | AY689125.1 | - | 2 - 3 | 267 | 90 | |
JN604984.1 | - | 3 - 4 | 745 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |