Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009263.2 | NP_001009263.1 | 4 - 5 | 389 | 95 | |
XM_012095263.3 | XP_011950653.1 | 4 - 5 | 381 | 95 | ||
GenBank mRNA | AJ401033.1 | - | 4 - 5 | 391 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |