Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAMBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009761.1 | NP_001009761.1 | 2 - 3 | 365 | 109 | |
GenBank mRNA | AY147883.1 | - | 2 - 3 | 365 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |