Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306114.1 | NP_001293043.1 | 2 - 3 | 241 | 65 | |
XM_012169456.3 | XP_012024846.2 | 3 - 4 | 416 | 65 | ||
XM_027965734.1 | XP_027821535.1 | 3 - 4 | 438 | 65 | ||
GenBank mRNA | KF055859.1 | - | 2 - 3 | 241 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |