Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306120.1 | NP_001293049.1 | 470 | 96 | ||
GenBank mRNA | JQ647512.1 | - | 393 | 96 | ||
KC290950.1 | - | 470 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |