Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306120.1 | NP_001293049.1 | 794 | 121 | ||
GenBank mRNA | JQ647512.1 | - | 717 | 121 | ||
KC290950.1 | - | 794 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |