Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163594.2 | NP_001157066.1 | 1 | 161 | 59 | |
GenBank mRNA | FJ937949.1 | - | 1 | 161 | 59 |
Gene Symbol: LOC114111037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962657.1 | XP_027818458.1 | 1 | 209 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |