Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RACK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281479.1 | NP_001268408.1 | 3 - 4 | 499 | 93 | |
GenBank mRNA | EU366471.1 | - | 1 - 2 | 164 | 93 | |
KC422451.1 | - | 3 - 4 | 499 | 93 |