Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162551.1 | NP_001156023.1 | 3 - 4 | 211 | 90 | |
GenBank mRNA | FJ969419.1 | - | 3 - 4 | 211 | 90 |