Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SQSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163043.1 | NP_001156515.1 | 3 - 4 | 536 | 79 | |
GenBank mRNA | FJ958359.1 | - | 3 - 4 | 536 | 79 |