Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245972.1 | NP_001232901.1 | 4 - 5 | 676 | 95 | |
GenBank mRNA | HQ263378.1 | - | 4 - 5 | 676 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |