Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134304.1 | NP_001127776.1 | 2 - 3 | 246 | 106 | |
GenBank mRNA | EU797607.1 | - | 2 - 3 | 246 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |