Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010053.4 | XP_004010102.3 | 9 - 10 | 2549 | 74 | |
XM_027970923.1 | XP_027826724.1 | 8 - 9 | 1578 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |