Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957162.1 | XP_027812963.1 | 6 - 7 | 3917 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |