Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011704.4 | XP_004011753.2 | 6 - 7 | 888 | 83 | |
XM_012183770.2 | XP_012039160.2 | 4 - 5 | 354 | 83 |